SureVector
Om denne vare
Combine standard DNA components to build custom vector constructs from Agilent’s set of standard parts.
- Rapid custom vector generation - Less than a day from design to vector, compared to 4 weeks for custom vector services
- Reliable and precise assembly - SureVector is extensively validated to ensure that standard parts can be interchanged, without loss of functionality
- More flexible than traditional systems - Assemble new vectors in your lab as experimental requirements change, versus always ordering a new one
- Control your experiments - Take control of your experiments, troubleshoot your own work, not that of your service providers DNA assembly
SureVector next-generation cloning uses the power of synthetic biology to combine standard DNA components to build custom vector constructs from the set of standard parts. Simply design a vector, order or PCR your gene-of-interest, and your custom plasmid is just a single reaction away.
Proprietary enzymes enable rapid and reliable assembly of multiple DNA fragments into a circular, recombinant plasmid containing your target gene.
Delivery information: Components include: bacterial selection markers, bacterial origins of replication, mammalian selection markers, promoters for yeast, mammalian cells and bacteria, and a variety of solubility/affinity tags; Enabling functionality in a range of cells.
Caution: For research use only. Not for use in diagnostic procedures.
3 tilgængelige muligheder nedenfor
Product Details & Documents
Combine standard DNA components to build custom vector constructs from Agilent’s set of standard parts.
- Rapid custom vector generation - Less than a day from design to vector, compared to 4 weeks for custom vector services
- Reliable and precise assembly - SureVector is extensively validated to ensure that standard parts can be interchanged, without loss of functionality
- More flexible than traditional systems - Assemble new vectors in your lab as experimental requirements change, versus always ordering a new one
- Control your experiments - Take control of your experiments, troubleshoot your own work, not that of your service providers DNA assembly
SureVector next-generation cloning uses the power of synthetic biology to combine standard DNA components to build custom vector constructs from the set of standard parts. Simply design a vector, order or PCR your gene-of-interest, and your custom plasmid is just a single reaction away.
Proprietary enzymes enable rapid and reliable assembly of multiple DNA fragments into a circular, recombinant plasmid containing your target gene.
Delivery information: Components include: bacterial selection markers, bacterial origins of replication, mammalian selection markers, promoters for yeast, mammalian cells and bacteria, and a variety of solubility/affinity tags; Enabling functionality in a range of cells.
Caution: For research use only. Not for use in diagnostic procedures.
Documents
SureVector Kit Ordering Guide
Additional Product Details
SureVector Next-Gen Cloning Kits The SureVector Cloning System
Application Notes
Product Family Options
Produktinformation
- PaknTilgængelighedPrice
- AGLSG7514ASureVector Core kit, SureVector Core contains the basic elements needed for functionality in bacterial, yeast, and mammalian systems, 15 reactionsSureVector Core kit, SureVector Core contains the basic elements needed for functionality in bacterial, yeast, and mammalian systems, 15 reactions1Log ind for tilgængelighed1 * 1 Items3.903,00 kr
- AGLSG7516BSureVector Mammalian C-terminal expansion kit, Promoters and C-terminal tags for expression in mammalian cells, 15 reactionsSureVector Mammalian C-terminal expansion kit, Promoters and C-terminal tags for expression in mammalian cells, 15 reactions1Log ind for tilgængelighed1 * 1 Items4.970,00 kr
- AGLSG7516ASureVector Mammalian N-terminal expansion kit, Promoters and N-terminal tags for expression in mammalian cells, 15 reactionsSureVector Mammalian N-terminal expansion kit, Promoters and N-terminal tags for expression in mammalian cells, 15 reactions1Log ind for tilgængelighed1 * 1 Items4.970,00 kr