Buckets, Round, with Lid
hvid, 2,1 L, 161/136 mm, 148 mm
Katalog # 216-0419
Marketsource
Leverandør: LAGAN PLAST EMBALLATOR
Gå til:
Product Details & Documents
PP, white.
- Material suitable for use with foodstuffs
- Sealed lid
- Good chemical and thermal resistance
Universal bucket for storage and transport, featuring a metal carrying handle.
Specifikationer
- Kapacitet:2,1 L
- Farve:hvid
- Højde:148 mm
- Ø top/ Ø bund:161/136 mm