Orders LinkContactUsLinkComponent
SPE cartridges, SPEC disk
SPE cartridges, SPEC disk
Leverandør:   VARIAN
SPE cartridges, SPEC disk
Leverandør:  VARIAN
Certifikater
SPE cartridges, SPEC disk
2 tilgængelige muligheder nedenfor
Product Details & Documents
SPEC Disk SPE delivers superior flow characteristics and trouble free automation.
  • No loose sorbent means no channeling and better reproducibility
  • Uniform flow and extraction properties offer robust performance
  • Low elution volume affords excellent concentration of the analyte, improving sensitivity
Bond Elut™ SPEC is a monolithic sorbent bed manufactured with ultraclean polypropylene plastic and bonded-silica impregnated on a glass fiber disc, resulting in an SPE product that delivers faster sample processing and lower costs. SPEC can result in increased productivity and lower extraction costs, without sacrificing the quality of the analytical results. For some applications, the elution volume is very small, meaning that the need to dry down and reconstitute the extract is eliminated. Integrated filtration protects against clogging.
Product Family Options
Produktinformation
  • Fase
    Kapacitet
    Lejevægt
    Tilgængelighed
    Price
  • VARIA5311120B
    Straight barrel cartridge
    Straight barrel cartridge
    MP1
    3 ml
    15 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 30.060,00 kr
    30.060,00 kr
    1 * 1.000 Items
       30.060,00 kr
  • VARIA5311120
    Straight barrel cartridge
    Straight barrel cartridge
    MP1
    3 ml
    15 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items - 3.250,00 kr
    3.250,00 kr
    1 * 100 Items
       3.250,00 kr