Orders LinkContactUsLinkComponent
Forensic anion solutions kit
Forensic anion solutions kit
Leverandør:   AGILENT HEWLETT PACKARD
Forensic anion solutions kit
Certifikater
Forensic anion solutions kit
2 tilgængelige muligheder nedenfor
Product Details & Documents
This highly focused kit was developed specifically for the analysis of poisonous compounds, such as cyanide, azide, selenate, arsenate, and arsenite.
  • Requires minimal sample preparation
In cases of poisoning, analytical tools are needed to determine the identity of the toxins quickly and accurately. A rapid determination of anionic toxins in adulterated foods and beverages is possible using CE with indirect UV detection. Forensic and other anions can be detected within 15 minutes with minimal sample preparation.
Product Family Options
Produktinformation
  • Pakn
    Tilgængelighed
    Price
  • HEWL5064-8208
    Forensic anion solutions kit for CE
    Forensic anion solutions kit for CE
    1
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 7.320,00 kr
    7.320,00 kr
    1 * 1 Items
       7.320,00 kr
  • HEWL5064-8209
    Buffer, basic anion, for CE, pH 12,1, 50 ml, indirect UV-detection buffer
    Buffer, basic anion, for CE, pH 12,1, 50 ml, indirect UV-detection buffer
    1
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 888,00 kr
    888,00 kr
    1 * 1 Items
       888,00 kr