GC columns, J&W PoraBOND U
Leverandør: VARIAN
Certifikater
Om denne vare
PoraBOND U is a highly stable polar-bonded porous polymer with the maximum temperature extended from 190 to 300 °C.
- Increased maximum temperature widens application range
- Reduced bleed delivers lower detection limits, more accurate results and faster stabilisation times
- Bonded PLOT phase for longevity
- Similar phases: Rt-U-BOND
Because the porous polymer is bonded to the column, the PoraBOND U is ideal for use with pressure programs, GC/MS applications and valve switching.
Frequently Bought Together