HRP - conjugate stabiliser, SeramunStab® STC
APOSBIL3115-100ML
:
- Pakn:100 mL
- Beskrivelse:SeramunStab® STC HRP - conjugate stabiliser
- Seq:01
Liquid product red odourless pH-value 7,2 to 7,4 boiling point 103 °C. Optimised for background reduction.