Orders LinkContactUsLinkComponent
Deoxynucleotide mix
Deoxynucleotide mix, PCR-grade, 0,4 ml, 1
Katalog # AGLS200415
Leverandør:   AGILENT
Deoxynucleotide mix
Katalog # AGLS200415
Leverandør:  AGILENT
CAS nummer:  
Deoxynucleotide mix
Deoxynucleotide mix, PCR-grade, 0,4 ml, 1
CAS nummer:  
Price
2.081,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The PCR grade deoxynucleotide mix includes 25 mM of each standard dNTP (GATC) for use with any PCR application, including real-time quantitative PCR.
  • Ideal for optimal PCR performance
  • Produces high-quality reaction products
  • Withstands multiple freeze-thaw without compromising efficiency
 : Includes 400 µl of 100 mM dNTP mix (25 mM of each dNTP), adequate for 1000 standard 50 µl primer-extension reactions or 2000 reactions of 25 µl.
Specifikationer
Specifikationer
  • Pakn:
    1
  • Beskrivelse:
    Deoxynucleotide mix, PCR-grade, 0,4 ml
Specifikationer