Om denne vare
- Diluted standard solutions that are ready-to-use for accurate measurements
- Traceable to SRM from NIST and USP
- Full certificates of analysis with comprehensive quality documentation
- Information on concentration and expanded measurement uncertainty
- Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Certifications: Traceable to SRM from NIST and USP.
8 tilgængelige muligheder nedenfor
Product Details & Documents
- Diluted standard solutions that are ready-to-use for accurate measurements
- Traceable to SRM from NIST and USP
- Full certificates of analysis with comprehensive quality documentation
- Information on concentration and expanded measurement uncertainty
- Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Certifications: Traceable to SRM from NIST and USP.
Documents
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- 1.25028.0100
- 1.25029.0100
- 1.25030.0100
- 1.25031.0100
- 1.25032.0100
- 1.25033.0100
- 1.25034.0100
- 1.25035.0100
- M-Clarity quality level:
- MQ200
- MQ200
- MQ200
- MQ200
- MQ200
- MQ200
- MQ200
- MQ200
Specifikationer
Product Family Options
Produktinformation
- KoncentrationNøjagtighedFör testkit(s)PakningpH-områdePaknTilgængelighedPrice
- 1.25028.0100COD standard solutionCOD standard solution20,0 mg/L± 0,7 mg/L1.14560; 1.01796Plastic bottle4 - 6100 mLLog ind for tilgængelighed1 * 100 mL383,00 krSpecifikationer:
More
Specifications
Cat. No.1.25028.0100M-Clarity quality levelMQ200 - 1.25029.0100COD standard solutionCOD standard solution100 mg/L± 3 mg/L1.14540; 1.14895; 1.14690; 1.14541Plastic bottle4 - 6100 mLLog ind for tilgængelighed1 * 100 mL383,00 krSpecifikationer:
More
Specifications
Cat. No.1.25029.0100M-Clarity quality levelMQ200 - 1.25030.0100COD standard solutionCOD standard solution200 mg/L± 4 mg/L1.14895; 1.14690; 1.14541Plastic bottle3 - 5100 mLLog ind for tilgængelighed1 * 100 mL383,00 krSpecifikationer:
More
Specifications
Cat. No.1.25030.0100M-Clarity quality levelMQ200 - 1.25031.0100COD standard solutionCOD standard solution400 mg/L± 5 mg/L1.14690; 1.14541; 1.14691Plastic bottle3 - 5100 mLLog ind for tilgængelighed1 * 100 mL428,00 krSpecifikationer:
More
Specifications
Cat. No.1.25031.0100M-Clarity quality levelMQ200 - 1.25032.0100COD standard solutionCOD standard solution1000 mg/L± 11 mg/L1.14541; 1.14691; 1.14555Plastic bottle3 - 5100 mLLog ind for tilgængelighed1 * 100 mL383,00 krSpecifikationer:
More
Specifications
Cat. No.1.25032.0100M-Clarity quality levelMQ200 - 1.25033.0100COD standard solutionCOD standard solution2000 mg/L± 32 mg/L1.14691; 1.14555Plastic bottle3 - 5100 mLLog ind for tilgængelighed1 * 100 mL384,00 krSpecifikationer:
More
Specifications
Cat. No.1.25033.0100M-Clarity quality levelMQ200 - 1.25034.0100COD standard solutionCOD standard solution8000 mg/L± 68 mg/L1.14555Plastic bottle3 - 5100 mLLog ind for tilgængelighed1 * 100 mL409,00 krSpecifikationer:
More
Specifications
Cat. No.1.25034.0100M-Clarity quality levelMQ200 - 1.25035.0100COD standard solutionCOD standard solution50000 mg/L± 894 mg/L1.01797Plastic bottle3 - 5100 mLLog ind for tilgængelighed1 * 100 mL462,00 krSpecifikationer:
More
Specifications
Cat. No.1.25035.0100M-Clarity quality levelMQ200