Coronavirus and SARS-CoV-2 ELISA kits, GSD NovaLisa®
Katalog # GSDCCVGQ0970
Supplier: GOLD STANDARD DIAGNOSTICS
CAS Number:
Specifications
- Pakn:96 Tests
- Beskrivelse:GSD NovaLisa® SARS-CoV-2 (COVID-19) quantitative IgG
- Seq:3
- Til:ThunderBolt® ELISA Analyser
- Test pr. sæt:1×96 reactions
Specifications
About this item
Immunoassay for the quantitative and qualitative detection of specific IgG-class antibodies to the spike protein of SARS-CoV-2.
- Quantitative determination of antibodies to the SARS-CoV-2 spike (S) protein
- Trimeric SARS-CoV-2 Spike antigen
- Intended as an aid to evaluate patients immune status and determine vaccine efficacy
- Calibrators correlate to First WHO International Standards for SARS-CoV-2
- Diagnostic sensitivity and specificity of 99.94% and 99.49% respectively
- Validated for serum, plasma, and finger prick (serum from capillary blood) samples
It is intended to aid in the diagnosis of patients suspected of having COVID-19 disease or asymptomatic presentation of SARS-CoV-2, to monitor antibody levels during and after a COVID-19 infection, and to determine antibody levels before and after a COVID-19 vaccination.