Orders LinkContactUsLinkComponent
VWR®, COD calibration standards, Certified Reference Materials (CRMs)
VWR®, COD calibration standards, Certified Reference Materials (CRMs)
Leverandør:  VWR Chemicals
Certifikater
VWR®, COD calibration standards, Certified Reference Materials (CRMs)
VWR®, COD calibration standards, Certified Reference Materials (CRMs)
17 tilgængelige muligheder nedenfor
Product Details & Documents
Avantor now offers a complete range of COD (Chemical Oxygen Demand) calibration standards (traceable to BAM) as CRM with concentrations between 20 and 10000 mg/L. Can be used to calibrate, control, verify or validate all COD test methods.
  • Traceable to BAM
  • Supplied with a detailed Certificate of Analysis
  • Complete range from 10 to 10000 mg/L
  • Customised COD standards available on request
  • Shelf life of 24 months / shelf life after opening: 1 month
  • Storage at 2 to 8 °C
  • Convenient pack size in 100 and 500 ml
COD is defined as the amount of oxygen equivalents consumed in the chemical oxidation of organic matter by strong oxidant (e.g., potassium dichromate) to carbon dioxide.
COD is often used as a measurement of pollutants in water, wastewater, and aqueous hazardous wastes.
The traditional chemical oxygen demand (COD) analysis method is the wet chemistry method. This involves a 2 hour digestion at high heat under acidic conditions in which potassium dichromate acts as the oxidant for any organic material present in a water sample. Silver sulphate is present as the catalyst and mercuric sulphate acts to complex out any interfering chloride. Following digestion, the extent of oxidation is measured through indirect measurement of oxygen demand via electrons consumed in the reduction of Cr⁶⁺ to Cr³⁺. This can be done by titration or spectrophotometry. Colorimetric measurement of COD is faster and easier method to perform than titrimetric analysis and does not require additional reagents.
COD is expressed in mg/L, which indicates the mass of oxygen consumed per litre of solution.
To ensure the lowest possible uncertainty levels COD calibration standards certified values are obtained by a weighted mean of the results of two independent calibration methods: Classical Volumetric and Instrumental (digestion of the chemically bonded organic carbon in the presence of a chemical oxidant and subsequent absorption measurement at 430 nm, using a spectrophotometer).
 : Certified Reference Materials acc. ISO 17025 and ISO 17034
Product Family Options
Produktinformation
  • Koncentration
    Pakning
    Pakn
    Tilgængelighed
    Price
  • 88406.260
    COD calibration standard
    COD calibration standard
    20 mg/L
    Glass bottle
    500 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 mL
    1 * 500 mL - 1.250,00 kr
    1.250,00 kr
    1 * 500 mL
       1.250,00 kr
  • 88407.180
    COD calibration standard
    COD calibration standard
    50 mg/L
    Glass bottle
    100 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mL
    1 * 100 mL - 851,00 kr
    851,00 kr
    1 * 100 mL
       851,00 kr
  • 88407.260
    COD calibration standard
    COD calibration standard
    50 mg/L
    Glass bottle
    500 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 mL
    1 * 500 mL - 1.250,00 kr
    1.250,00 kr
    1 * 500 mL
       1.250,00 kr
  • 88408.180
    COD calibration standard
    COD calibration standard
    100 mg/L
    Glass bottle
    100 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mL
    1 * 100 mL - 851,00 kr
    851,00 kr
    1 * 100 mL
       851,00 kr
  • 88408.260
    COD calibration standard
    COD calibration standard
    100 mg/L
    Glass bottle
    500 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 mL
    1 * 500 mL - 1.250,00 kr
    1.250,00 kr
    1 * 500 mL
       1.250,00 kr
  • 88409.180
    COD calibration standard
    COD calibration standard
    200 mg/L
    Glass bottle
    100 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mL
    1 * 100 mL - 851,00 kr
    851,00 kr
    1 * 100 mL
       851,00 kr
  • 88409.260
    COD calibration standard
    COD calibration standard
    200 mg/L
    Glass bottle
    500 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 mL
    1 * 500 mL - 1.250,00 kr
    1.250,00 kr
    1 * 500 mL
       1.250,00 kr
  • 88410.180
    COD calibration standard
    COD calibration standard
    400 mg/L
    Glass bottle
    100 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mL
    1 * 100 mL - 851,00 kr
    851,00 kr
    1 * 100 mL
       851,00 kr
  • 88410.260
    COD calibration standard
    COD calibration standard
    400 mg/L
    Glass bottle
    500 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 mL
    1 * 500 mL - 1.250,00 kr
    1.250,00 kr
    1 * 500 mL
       1.250,00 kr
  • 88411.180
    COD calibration standard
    COD calibration standard
    500 mg/L
    Glass bottle
    100 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mL
    1 * 100 mL - 851,00 kr
    851,00 kr
    1 * 100 mL
       851,00 kr
  • 88411.260
    COD calibration standard
    COD calibration standard
    500 mg/L
    Glass bottle
    500 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 mL
    1 * 500 mL - 1.250,00 kr
    1.250,00 kr
    1 * 500 mL
       1.250,00 kr
  • 88412.180
    COD calibration standard
    COD calibration standard
    1000 mg/L
    Glass bottle
    100 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mL
    1 * 100 mL - 851,00 kr
    851,00 kr
    1 * 100 mL
       851,00 kr
  • 88412.260
    COD calibration standard
    COD calibration standard
    1000 mg/L
    Glass bottle
    500 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 mL
    1 * 500 mL - 1.250,00 kr
    1.250,00 kr
    1 * 500 mL
       1.250,00 kr
  • 88413.180
    COD calibration standard
    COD calibration standard
    2000 mg/L
    Glass bottle
    100 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mL
    1 * 100 mL - 851,00 kr
    851,00 kr
    1 * 100 mL
       851,00 kr
  • 88413.260
    COD calibration standard
    COD calibration standard
    2000 mg/L
    Glass bottle
    500 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 mL
    1 * 500 mL - 1.250,00 kr
    1.250,00 kr
    1 * 500 mL
       1.250,00 kr
  • 88414.180
    COD calibration standard
    COD calibration standard
    10000 mg/L
    Glass bottle
    100 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mL
    1 * 100 mL - 993,00 kr
    993,00 kr
    1 * 100 mL
       993,00 kr
  • 88414.260
    COD calibration standard
    COD calibration standard
    10000 mg/L
    Glass bottle
    500 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 mL
    1 * 500 mL - 1.680,00 kr
    1.680,00 kr
    1 * 500 mL
       1.680,00 kr