Orders LinkContactUsLinkComponent
b.safe Funnels 180
b.safe Funnels 180
Leverandør:   Bohlender
b.safe Funnels 180
Leverandør:  Bohlender
Certifikater
b.safe Funnels 180
24 tilgængelige muligheder nedenfor
Product Details & Documents
Funnel made of PE / static dissipative PE-EX for liquid waste disposal. The funnel can be safely mounted on the waste container with its freely turnable cap that is safely fixed with the filling neck.
  • Easy and safe handling: you do not need to open a lid before pouring
  • Removable sieve restrains, e.g. magnetic stirring bars or other solids
  • Splash guard for a smooth flow without splashing
  • Ball valve protects from harmful vapours; it floats up during filling and opens the drain
  • Free turnable cap for easy assembly on collecting container
  • Neck with plastic nibs to fix attachable filling tubes for under level filling
b.safe Funnels 180 are available in many versions. Choose the right one for your application: The sieve restrains magnetic stirrer bars and other solid materials and can be easily removed for cleaning. The hinged lid respectively the ball valve reliably withhold vapours from the container. The splash guard avoids spillover and splashing during funnel filling. The static dissipative PE-EX version includes two attachable filling tubes (Ø 36 x usable length 97 mm per tube) as well as an 1.5 m long grounding cable.
Safe earthing of the funnel can be made via the grounding cable by qualified personnel. The mounted filling tube allows an under level filling to avoid static charging.
Documents
Product Family Options
Produktinformation
  • Gevind
    Tilgængelighed
    Price
  • 590-0362
    Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    GL45
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.560,00 kr
    1.560,00 kr
    1 * 1 Items
       1.560,00 kr
  • 590-0366
    Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    S50
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
  • 590-0371
    Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    S51
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.580,00 kr
    1.580,00 kr
    1 * 1 Items
       1.580,00 kr
  • 590-0377
    Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    S55
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.610,00 kr
    1.610,00 kr
    1 * 1 Items
       1.610,00 kr
  • 590-0387
    Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    S60
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.580,00 kr
    1.580,00 kr
    1 * 1 Items
       1.580,00 kr
  • 590-0393
    Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    S65
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.930,00 kr
    1.930,00 kr
    1 * 1 Items
       1.930,00 kr
  • 590-0397
    Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    S70
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.650,00 kr
    1.650,00 kr
    1 * 1 Items
       1.650,00 kr
  • 590-0400
    Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Funnel with sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    S90
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.650,00 kr
    1.650,00 kr
    1 * 1 Items
       1.650,00 kr
  • 590-0360
    Static dissipative funnel with grounding cable, sieve and lid
    Static dissipative funnel with grounding cable, sieve and lid
    GL45
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.280,00 kr
    2.280,00 kr
    1 * 1 Items
       2.280,00 kr
  • 590-0374
    Static dissipative funnel with grounding cable, sieve and lid
    Static dissipative funnel with grounding cable, sieve and lid
    S55
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.040,00 kr
    2.040,00 kr
    1 * 1 Items
       2.040,00 kr
  • 590-0380
    Static dissipative funnel with grounding cable, sieve and lid
    Static dissipative funnel with grounding cable, sieve and lid
    S60
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.220,00 kr
    2.220,00 kr
    1 * 1 Items
       2.220,00 kr
  • 590-0402
    Static dissipative funnel with grounding cable, sieve and lid
    Static dissipative funnel with grounding cable, sieve and lid
    G2"+Trisure2"
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 3.250,00 kr
    3.250,00 kr
    1 * 1 Items
       3.250,00 kr
  • 590-0406
    Static dissipative funnel with grounding cable, sieve and lid
    Static dissipative funnel with grounding cable, sieve and lid
    Mauser 2"
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.920,00 kr
    2.920,00 kr
    1 * 1 Items
       2.920,00 kr
  • 590-0358
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve
    GL45
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.560,00 kr
    1.560,00 kr
    1 * 1 Items
       1.560,00 kr
  • 590-0359
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    GL45
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.190,00 kr
    2.190,00 kr
    1 * 1 Items
       2.190,00 kr
  • 590-0364
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    S50
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.270,00 kr
    2.270,00 kr
    1 * 1 Items
       2.270,00 kr
  • 590-0368
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    S51
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.180,00 kr
    2.180,00 kr
    1 * 1 Items
       2.180,00 kr
  • 590-0373
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    S55
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.200,00 kr
    2.200,00 kr
    1 * 1 Items
       2.200,00 kr
  • 590-0379
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    S60
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.160,00 kr
    2.160,00 kr
    1 * 1 Items
       2.160,00 kr
  • 590-0390
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    S65
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.170,00 kr
    2.170,00 kr
    1 * 1 Items
       2.170,00 kr
  • 590-0395
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    S70
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.570,00 kr
    2.570,00 kr
    1 * 1 Items
       2.570,00 kr
  • 590-0399
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    S90
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.510,00 kr
    2.510,00 kr
    1 * 1 Items
       2.510,00 kr
  • 590-0401
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    G2"+Trisure2"
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.790,00 kr
    2.790,00 kr
    1 * 1 Items
       2.790,00 kr
  • 590-0405
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Static dissipative funnel with grounding cable, sieve, splash guard, ball valve and 2× Ø 36 mm filling tubes
    Mauser 2"
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.350,00 kr
    2.350,00 kr
    1 * 1 Items
       2.350,00 kr
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Static dissipative filling tubes for funnel 180
    Static dissipative filling tubes for funnel 180
    590-0319
    Log ind for tilgængelighed
    1 * 5 Items - 605,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Static dissipative sieve for funnel 180
    Static dissipative sieve for funnel 180
    590-0311
    Log ind for tilgængelighed
    1 * 1 Items - 346,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Sieve for funnel 180
    Sieve for funnel 180
    590-0312
    Log ind for tilgængelighed
    1 * 1 Items - 298,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Static dissipative splash guard for funnel 180 without hinged lid
    Static dissipative splash guard for funnel 180 without hinged lid
    590-0315
    Log ind for tilgængelighed
    1 * 1 Items - 316,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Splash guard for funnel 180 without hinged lid
    Splash guard for funnel 180 without hinged lid
    590-0316
    Log ind for tilgængelighed
    1 * 1 Items - 281,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Filling tubes for funnel 180
    Filling tubes for funnel 180
    590-0320
    Log ind for tilgængelighed
    1 * 5 Items - 189,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Static dissipative angled adaptor for canisters, for vertical adjustment of b.safe Funnels 130 with thread S60 on b.safe canisters S60 with inclined neck
    Static dissipative angled adaptor for canisters, for vertical adjustment of b.safe Funnels 130 with thread S60 on b.safe canisters S60 with inclined neck
    590-0333
    Log ind for tilgængelighed
    1 * 1 Items - 1.190,00 kr