Fluorbenzen 99%
Bottle, 100 g
Katalog # APOSPC3480-100G
Leverandør: Apollo Scientific
Gå til:
Product Details & Documents
Alternative solvent to benzene which is less toxic.
Synonymer: Phenylfluoride
- CAS nummer:462-06-6
- Formel:C₆H₅F
- MDL nummer:MFCD00000280
- Molecular Weight:96.1 g/mol
- Kogende Pt:84…85 °C
- Smeltende Pt:–42 °C
- Tæthed:1,024 g/cm³ (25 °C)
- Flash Pt:–12 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:207-321-7
- FN:2387
- ADR:3,II
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:100 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



