4-Ethylanisol 98%
Leverandør: Apollo Scientific
Gå til:
Om denne vare
Synonymer: 1-Ethyl-4-methoxybenzene
- CAS nummer:1515-95-3
- Formel:C₉H₁₂O
- MDL nummer:MFCD00015176
- Molecular Weight:136.19 g/mol
- Opbevaringstemperatur:Stue temperatur
6 tilgængelige muligheder nedenfor
Product Details & Documents
Synonymer: 1-Ethyl-4-methoxybenzene
- CAS nummer:1515-95-3
- Formel:C₉H₁₂O
- MDL nummer:MFCD00015176
- Molecular Weight:136.19 g/mol
- Opbevaringstemperatur:Stue temperatur
Product Family Options
Produktinformation
- PakningPaknTilgængelighedPrice
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



