Orders LinkContactUsLinkComponent
5-Bromisoquinolin 98%
5-Bromisoquinolin 98%
Irritant
Supplier:   Apollo Scientific
5-Bromisoquinolin 98%
Supplier:  Apollo Scientific
Certificates
5-Bromisoquinolin 98%
3 Options Available Below
Product Details & Documents
  • CAS Number:34784-04-8
  • Formula:C₉H₆BrN
  • MDL Number:MFCD01646405
  • Molecular Weight:208.06 g/mol
  • Boiling Pt:95…97 °C/0,1mmHg
  • Melting Pt:81…83 °C
  • Storage Temperature:Stue temperatur
Product Family Options
Product Information
  • Pakning
    Pakn
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 165,00 kr
    165,00 kr
    1 * 5 g
       165,00 kr
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 527,00 kr
    527,00 kr
    1 * 25 g
       527,00 kr
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 1.660,00 kr
    1.660,00 kr
    1 * 100 g
       1.660,00 kr