Orders LinkContactUsLinkComponent
3'-Chlorpropiophenon
Bottle, 25 g
Catalog # APOSOR8786-25G
Irritant
Supplier:   Apollo Scientific
3'-Chlorpropiophenon
Catalog # APOSOR8786-25G
Supplier:  Apollo Scientific
CAS Number:  34841-35-5
3'-Chlorpropiophenon
Bottle, 25 g
CAS Number:  34841-35-5
Price
165,00 kr
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  m-Chloropropiophenone, 1-(3-Chlorophenyl)propan-1-one
  • CAS Number:34841-35-5
  • Formula:C₉H₉ClO
  • MDL Number:MFCD00009925
  • Molecular Weight:168.62 g/mol
  • Boiling Pt:123…124 °C/13mmHg
  • Melting Pt:45…47 °C
  • Flash Pt:>110 °C
  • Storage Temperature:Stue temperatur
  • EINECS:252-242-3
Specifications
Specifications
  • Pakning:
    Bottle
  • Pakn:
    25 g