Orders LinkContactUsLinkComponent
7-Methylindol
7-Methylindol
Irritant
Supplier:   Apollo Scientific
7-Methylindol
Supplier:  Apollo Scientific
Certificates
3 Options Available Below
Product Details & Documents
  • CAS Number:933-67-5
  • Formula:C₉H₉N
  • MDL Number:MFCD00005684
  • Molecular Weight:131.18 g/mol
  • Boiling Pt:266 °C
  • Melting Pt:80…84 °C
  •  :Density: 1.0202 g/cm³ (99.6 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:213-270-1
Product Family Options
Product Information
  • Pakning
    Pakn
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 428,00 kr
    428,00 kr
    1 * 25 g
       428,00 kr
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 1.680,00 kr
    1.680,00 kr
    1 * 100 g
       1.680,00 kr
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 8.220,00 kr
    8.220,00 kr
    1 * 500 g
       8.220,00 kr
    Specifications:
    New Product