5-Methoxyoxindole
Leverandør: Apollo Scientific
Gå til:
Om denne vare
Synonymer: 5-Methoxy-2-indolinone, 5-Methoxyindolin-2-one
- CAS nummer:7699-18-5
- Formel:C9H9NO2
- MDL nummer:MFCD01860251
- Molecular Weight:163.1733 g/mol
- Opbevaringstemperatur:Køleskab
4 tilgængelige muligheder nedenfor
Product Details & Documents
Synonymer: 5-Methoxy-2-indolinone, 5-Methoxyindolin-2-one
- CAS nummer:7699-18-5
- Formel:C9H9NO2
- MDL nummer:MFCD01860251
- Molecular Weight:163.1733 g/mol
- Opbevaringstemperatur:Køleskab
Product Family Options
Produktinformation
- PakningPaknTilgængelighedPrice
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
- Specifikationer:New ProductNew Product
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



