Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Methylblå

Methylblå

Bottle
Catalog # APOSBIA4014-25G
New Product
Supplier:   Apollo Scientific
Methylblå
Catalog # APOSBIA4014-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  28983-56-4
Methylblå

Bottle

CAS Number:  28983-56-4
Price
275,00 kr
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  C.I. 42780
  • CAS Number:28983-56-4
  • Formula:C₃₇H₂₇N₃Na₂O₉S₃
  • MDL Number:MFCD00058509
  • Molecular Weight:799.81 g/mol
  • Storage Temperature:Stue temperatur
Specifications

Specifications

  • Pakning:
    Bottle

Specifications