SafetyCaps
Om denne vare
The SCAT SafetyCaps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapours. Complete with integrated air valve for pressure equalisation.
- High quality materials ensure maximum safety for lab and staff
- 360° rotatable and up to 200 °C autoclavable screw cap for maximum flexibility
- Stable construction for highest safety and easy handling
- Chemically resistant against aggressive organic solvents
- Attractive design and clear arrangement of the connections
- Improved design for easy connection of capillaries in various colours
- Free air vaive with change indicator included with every SafetyCap
SCAT SafetyCaps are available in the following versions:
SafetyCaps, basic: Standard SafetyCap with different connections without additional functions.
SafetyCaps, preparative: For use in the preparative HPLC environment with larger tubing connections and air valves with a higher flow rate.
SafetyCaps, with shut-off: Prevent air pockets in the tube when the supply bottle is being changed and guarantees that the analysis continues without a problem.
SafetyCaps, combined: Provides a stable baseline and reliable reproducibility for RI detection for a fast and simple solvent degassing with helium.
SafetyCaps, angled: With lateral connections for capillary and air valve in case of limited space above the bottle.
SafetyCaps, for Merck® bottles: Connect containers with thread sizes S 40 and GL 40 directly to the HPLC system, without decanting or thread adapters.
SafetyCaps, for NS 29/32 ground neck bottles: The locknut allows the stopper to be easily loosened and removed for bottles with NS 29/32 mm standard ground joints.
Delivery information: All SafetyCaps are delivered with an air valve and corresponding fittings. PFA fittings, PTFE body and PPS screw cap are conforming to the requirements of flammability classification V-0 (UL-94). Blind plugs are provided to tighten connections which are temporarily inactive or not in use.
21 tilgængelige muligheder nedenfor
Product Details & Documents
The SCAT SafetyCaps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapours. Complete with integrated air valve for pressure equalisation.
- High quality materials ensure maximum safety for lab and staff
- 360° rotatable and up to 200 °C autoclavable screw cap for maximum flexibility
- Stable construction for highest safety and easy handling
- Chemically resistant against aggressive organic solvents
- Attractive design and clear arrangement of the connections
- Improved design for easy connection of capillaries in various colours
- Free air vaive with change indicator included with every SafetyCap
SCAT SafetyCaps are available in the following versions:
SafetyCaps, basic: Standard SafetyCap with different connections without additional functions.
SafetyCaps, preparative: For use in the preparative HPLC environment with larger tubing connections and air valves with a higher flow rate.
SafetyCaps, with shut-off: Prevent air pockets in the tube when the supply bottle is being changed and guarantees that the analysis continues without a problem.
SafetyCaps, combined: Provides a stable baseline and reliable reproducibility for RI detection for a fast and simple solvent degassing with helium.
SafetyCaps, angled: With lateral connections for capillary and air valve in case of limited space above the bottle.
SafetyCaps, for Merck® bottles: Connect containers with thread sizes S 40 and GL 40 directly to the HPLC system, without decanting or thread adapters.
SafetyCaps, for NS 29/32 ground neck bottles: The locknut allows the stopper to be easily loosened and removed for bottles with NS 29/32 mm standard ground joints.
Delivery information: All SafetyCaps are delivered with an air valve and corresponding fittings. PFA fittings, PTFE body and PPS screw cap are conforming to the requirements of flammability classification V-0 (UL-94). Blind plugs are provided to tighten connections which are temporarily inactive or not in use.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Product Family Options
Basic
Produktinformation
- GevindStikTilgængelighedPrice
- 590-0521SafetyCap IV
- 590-0516SafetyCap I
- 590-0517SafetyCap II
- 590-0518SafetyCap III
- 221-0262SafetyCap II
- 554-4688SafetyCap IIISafetyCap IIIGL383× PFA-fitting 1,6/2,3/3,2 mm OD, 2× PFA blind plugLog ind for tilgængelighed1 * 1 Items886,00 kr
- SCTV107511SafetyCap II
- SCTV307019SafetyCap I, V2.0SafetyCap I, V2.0GL451× PFA-fitting 1,6/2,3/3,2 mm ODLog ind for tilgængelighed1 * 1 Items753,00 kr
- SCTV307006SafetyCap IISafetyCap IIGL282× PFA-fitting 1,6/2,3/3,2 mm OD, 1× PFA-blind plugLog ind for tilgængelighed1 * 1 Items886,00 kr
- SCTV307410SafetyCap IV, V2.0SafetyCap IV, V2.0GL454× PFA-fitting 1,6/2,3/3,2 mm ODLog ind for tilgængelighed1 * 1 Items1.070,00 kr
- SCTV307520SafetyCap VI, V2.0SafetyCap VI, V2.0GL456× PFA-fitting 1,6/2,3/3,2 mm ODLog ind for tilgængelighed1 * 1 Items1.060,00 kr
- SCTV307909SafetyCap II, V2.0SafetyCap II, V2.0GL452× PFA-fitting 1,6/2,3/3,2 mm ODLog ind for tilgængelighed1 * 1 Items906,00 kr
- SCTV307910SafetyCap III, V2.0SafetyCap III, V2.0GL453× PFA-fitting 1,6/2,3/3,2 mm ODLog ind for tilgængelighed1 * 1 Items886,00 kr
Angled
Produktinformation
- GevindStikTilgængelighedPrice
- SCTV399019SafetyCap I, V2.0, 90° angledSafetyCap I, V2.0, 90° angledGL451× PFA-fitting 3,2 mm ODLog ind for tilgængelighed1 * 1 Items1.060,00 kr
- SCTV399909SafetyCap II, V2.0, 90° angledSafetyCap II, V2.0, 90° angledGL452× PFA-fitting 3,2 mm ODLog ind for tilgængelighed1 * 1 Items1.150,00 kr
For special bottles
Produktinformation
- GevindStikTilgængelighedPrice
- 590-0553SafetyCap II, for ground neck bottles 29/32 mm, with locknutSafetyCap II, for ground neck bottles 29/32 mm, with locknut29/32 mm2× PFA-fitting 3,2 mm ODLog ind for tilgængelighed1 * 1 Items1.060,00 kr
- 554-3494SafetyCap I, for Merck® bottles with shut-offSafetyCap I, for Merck® bottles with shut-offS401× PFA-fitting 3,2 mm ODLog ind for tilgængelighed1 * 1 Items1.150,00 kr
- SCTV107637Safety Cap II for Wheaton bottles
- SCTV107742SafetyCap III, for Merck® bottlesSafetyCap III, for Merck® bottlesS403× PFA-fitting 3,2 mm ODLog ind for tilgængelighed1 * 1 Items966,00 kr
- SCTV307100SafetyCap I, V2.0, for Merck® bottlesSafetyCap I, V2.0, for Merck® bottlesGL/S401× PFA-fitting 1,6/2,3/3,2 mm ODLog ind for tilgængelighed1 * 1 Items797,00 kr
- SCTV307101SafetyCap II, V2.0, for Merck® bottlesSafetyCap II, V2.0, for Merck® bottlesGL/S402× PFA-fitting 1,6/2,3/3,2 mm OD, 1× PFA-blind plugLog ind for tilgængelighed1 * 1 Items886,00 kr
Preperative
Produktinformation
- Katalog #GevindStikBeskrivelseTilgængelighedPrice
- GL452× PTFE-fitting 6,35 mm ODSafetyCap II, V2.0, preperativeLog ind for tilgængelighed1 * 1 Items966,00 kr
With shut-off
Produktinformation
- Katalog #GevindStikBeskrivelseTilgængelighedPrice
- GL454× PFA-fittings 3,2 mm ODSafetyCap IV, V2.0, with 4× shut-offLog ind for tilgængelighed1 * 1 Items1.330,00 kr
Replacement air valves
Produktinformation
- Katalog #BeskrivelseTilgængelighedPrice
- Air valve for SafetyCaps V 2.0, UNF ¹/₄" 28G, service life 6 monthsLog ind for tilgængelighed1 * 1 Items221,00 kr
Replacement PFA fittings
Produktinformation
- Katalog #FarveBeskrivelseMaterialeFor tubing ext.ØTilgængelighedPrice