Filling support for 10 L compact jerrycans
Catalog # 331-0380
Supplier: BURKLE
Specifications
- Til:10 L jerricans
- Beskrivelse:Filling support
- Pakn:1
Specifications
About this item
The filling support makes it easy to dispense liquids and correctly dose from Bürkle's 10 litre compact jerrycan.
- Stainless steel V2A (1.4301)
- For use with Bürkle compact jerrycans 10 litres (PP, transparent or HDPE, black, antistatic)
The jerrycan can be reliably guided with the swivelling holder and the liquid medium can be dispensed and transferred without expending any great effort.