Orders LinkContactUsLinkComponent
Safety canisters
Safety canisters
Leverandør:   S.C.A.T.
Safety canisters
Leverandør:  S.C.A.T.
Certifikater
Safety canisters
Safety canisters
18 tilgængelige muligheder nedenfor
Product Details & Documents
These canisters are made of high-quality PE-HD material with high chemical resistance for use in the laboratory for safe disposal of chromatographic solvents or other liquid laboratory waste for the highest level of work safety in the laboratory.
  • Made of very robust HDPE, which is very resistant to almost all solvents commonly used in the laboratory
  • Smallest canisters have a volume of only 5 L and are ideally suited for use on tables or smaller storage areas
  • Larger canisters are available with volumes of 10, 12, 20, 30 up to 60 L, which can be deposited under tables or in safety cabinets
  • Thread types S55, S60/61, S65, S70/71, S90 and S95 are commonly used in the laboratory, they are all compatible with the SCAT SafetyWasteCaps range for the safe disposal of volatile hazardous substances
  • Special versions available with floats for level indication or with internal fluorination for double protection of the contents, both for permeation from outside to inside and for diffusion from inside to outside
The wide range of non-electrically conductive canisters includes a variety of designs for individual use in the laboratory for the disposal of solvents in combination with the SCAT SafetyWasteCaps to increase your laboratory safety.
Most SCAT canisters are UN-Y or UN-X approved, which ensures safe storage or safe transport of eluents.
Product Family Options
UN-certified
Produktinformation
  • Kapacitet
    Farve
    Gevind
    Model
    B×D×H
    Tilgængelighed
    Price
  • 590-0647
    Safety canister
    Safety canister
    2,5 L
    ufarvet
    GL 45
    UN-Y approval
    115×150×210 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 136,00 kr
    136,00 kr
    1 * 1 Items
       136,00 kr
  • 590-0648
    Safety canister
    Safety canister
    5 L
    ufarvet
    GL 45
    UN-Y approval
    150×195×250 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 181,00 kr
    181,00 kr
    1 * 1 Items
       181,00 kr
  • 590-0655
    Safety canister
    Safety canister
    5 L
    ufarvet
    S 51
    UN-Y approval
    145×190×250 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 177,00 kr
    177,00 kr
    1 * 1 Items
       177,00 kr
  • SCTV107933
    Safety canister
    Safety canister
    5 L
    ufarvet
    S 55
    UN-X approval; with inner fluorination
    145×190×250 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 221,00 kr
    221,00 kr
    1 * 1 Items
       221,00 kr
  • 590-0654
    Safety canister
    Safety canister
    5 L
    ufarvet
    S 55
    UN-Y approval
    160×185×230 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 193,00 kr
    193,00 kr
    1 * 1 Items
       193,00 kr
  • SCTV108175
    Safety canister
    Safety canister
    5 L
    ufarvet
    S 55
    UN-X approval
    182×162×240 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 177,00 kr
    177,00 kr
    1 * 1 Items
       177,00 kr
  • SCTV108227
    Safety canister
    Safety canister
    5 L
    ufarvet
    S 60/61
    UN-Y approval
    164×185×235 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 177,00 kr
    177,00 kr
    1 * 1 Items
       177,00 kr
  • 590-0574
    Sikkerhedsbeholder
    Sikkerhedsbeholder
    5 L
    ufarvet
    S 95
    UN-Y approval
    170×170×310 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 177,00 kr
    177,00 kr
    1 * 1 Items
       177,00 kr
  • 590-0649
    Safety canister
    Safety canister
    10 L
    ufarvet
    GL 45
    UN-Y approval
    190×230×305 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 195,00 kr
    195,00 kr
    1 * 1 Items
       195,00 kr
  • 590-0652
    Safety canister
    Safety canister
    10 L
    ufarvet
    S 55
    UN-Y approval
    185×225×305 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 177,00 kr
    177,00 kr
    1 * 1 Items
       177,00 kr
  • SCTV108189
    Safety canister
    Safety canister
    10 L
    ufarvet
    S 55
    UN-Y approval, with inner fluorination
    190×230×310 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 443,00 kr
    443,00 kr
    1 * 1 Items
       443,00 kr
  • 590-0710
    Safety canister
    Safety canister
    10 L
    ufarvet
    S 90
    UN-Y approval
    195×195×380 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 265,00 kr
    265,00 kr
    1 * 1 Items
       265,00 kr
  • 554-1849
    Safety canister
    Safety canister
    12 L
    ufarvet
    S 60/61
    UN-Y approval
    200×235×350 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 195,00 kr
    195,00 kr
    1 * 1 Items
       195,00 kr
  • 590-0653
    Sikkerhedsbeholder
    Sikkerhedsbeholder
    20 L
    ufarvet
    S 60/61
    UN-Y approval
    260×289×390 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 221,00 kr
    221,00 kr
    1 * 1 Items
       221,00 kr
  • 590-0656
    Sikkerhedsbeholder
    Sikkerhedsbeholder
    30 L
    ufarvet
    S 60/61
    UN-Y approval
    290×380×400 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 265,00 kr
    265,00 kr
    1 * 1 Items
       265,00 kr
  • SCTV107710
    Safety canister
    Safety canister
    60 L
    ufarvet
    S 70/71
    UN-Y approval
    330×370×635 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 708,00 kr
    708,00 kr
    1 * 1 Items
       708,00 kr
Non UN-certified
Produktinformation
  • Kapacitet
    Farve
    Gevind
    Model
    B×D×H
    Tilgængelighed
    Price
  • 590-0741
    Sikkerhedsbeholder
    Sikkerhedsbeholder
    20 L
    ufarvet
    S 60/61
    With floater
    260×285×455 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 966,00 kr
    966,00 kr
    1 * 1 Items
       966,00 kr
  • SCTV108115
    Safety canister
    Safety canister
    30 L
    ufarvet
    S 60/61 and GL 45
    2 openings
    290×380×400 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.240,00 kr
    1.240,00 kr
    1 * 1 Items
       1.240,00 kr
Replacement lid
Produktinformation
  • Katalog #
    Farve
    Gevind
    Beskrivelse
    Tilgængelighed
    Price
  • sort
    S 70/71
    Screw cap for canister SCTV107710
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 34,60 kr
    34,60 kr
    1 * 1 Items
       34,60 kr