Gloves, latex, Flex Grip, A174
Supplier: Portwest
Certificates
About this item
Flex Grip gloves designed using a seamless knitted liner, with a waterproof latex coated palm, this glove promotes a high level of comfort, flexibility and softness.
- Latex foam coating for excellent grip in wet and dry conditions
- 13 gauge liner for a perfect fit
- Elasticated cuffs for a secure fit
The ergonomic design mimics the hands natural curvature reducing hand fatigue. Breathable open back of hand aids to reduce perspiration.
Colour: Red/Black
Certifications: EN 420, EN 388:2016 +A1:2018 (2.1.2.1.X), AS/NZS 2161 .2, AS/NZS 2161 .3
Specifications
- Catalog No:
- PRTWA174R8RS
- PRTWA174R8RXXL
- PRTWA174R8RXL
- PRTWA174R8RM
- PRTWA174R8RL
- Model:
- Flex Grip A174
- Flex Grip A174
- Flex Grip A174
- Flex Grip A174
- Flex Grip A174
- Ambidextrous/Hånd-specifik:
- Hånd-specifik
- Hånd-specifik
- Hånd-specifik
- Hånd-specifik
- Hånd-specifik
- Engangs/Genbrugelig:
- Genanvendelig
- Genanvendelig
- Genanvendelig
- Genanvendelig
- Genanvendelig
- Foringsmateriale:
- Strikke
- Strikke
- Strikke
- Strikke
- Strikke
- Materiale:
- Latex
- Latex
- Latex
- Latex
- Latex
- Overflade:
- Latex coating on palm and fingertips
- Latex coating on palm and fingertips
- Latex coating on palm and fingertips
- Latex coating on palm and fingertips
- Latex coating on palm and fingertips
- Måler:
- 13
- 13
- 13
- 13
- 13
- Manchet:
- Strikket
- Strikket
- Strikket
- Strikket
- Strikket
- EN norm:
- EN 420, EN 388 (2.1.2.1.X)
- EN 420, EN 388 (2.1.2.1.X)
- EN 420, EN 388 (2.1.2.1.X)
- EN 420, EN 388 (2.1.2.1.X)
- EN 420, EN 388 (2.1.2.1.X)