High visibility softshell jackets, S424
Om denne vare
Classically styled, gives enhanced protection whatever the elements. With a mobile phone pocket and a radio loop as standard, this jacket is warm, comfortable and water repellent.
- Micro polar fleece lined and bonded with shell
- Reflective tape for increased visibility
- Four pockets for ample storage
- Two side zip pockets for secure storage
- Zipped chest pocket and internal pocket for safe storage
- Hook and loop cuffs for a secure fit
- Radio loop for easy clipping of a radio
Features include breathable fabric to draw moisture away from the body keeping the wearer cool, dry and comfortable. Extremely water resistant fabric finish, water beads away from fabric surface.
Shell fabric: 94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar lleece 150 g, 310 g.
Certifications: EN ISO 20471 Class 3; AS/NZS 4602.1:2011 Class D/N; RIS-3279-TOM ISSUE 1 (orange only); GO/RT 3279 Issue 8 (orange only); CE
12 tilgængelige muligheder nedenfor
Product Details & Documents
Classically styled, gives enhanced protection whatever the elements. With a mobile phone pocket and a radio loop as standard, this jacket is warm, comfortable and water repellent.
- Micro polar fleece lined and bonded with shell
- Reflective tape for increased visibility
- Four pockets for ample storage
- Two side zip pockets for secure storage
- Zipped chest pocket and internal pocket for safe storage
- Hook and loop cuffs for a secure fit
- Radio loop for easy clipping of a radio
Features include breathable fabric to draw moisture away from the body keeping the wearer cool, dry and comfortable. Extremely water resistant fabric finish, water beads away from fabric surface.
Shell fabric: 94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar lleece 150 g, 310 g.
Certifications: EN ISO 20471 Class 3; AS/NZS 4602.1:2011 Class D/N; RIS-3279-TOM ISSUE 1 (orange only); GO/RT 3279 Issue 8 (orange only); CE
Specifications for Product Family
Specifikationer
- Catalog No:
- PRTWS424ORRS
- PRTWS424ORRM
- PRTWS424ORRL
- PRTWS424ORRXL
- PRTWS424ORRXXL
- PRTWS424ORRXXXL
- PRTWS424YERS
- PRTWS424YERM
- PRTWS424YERL
- PRTWS424YERXL
- PRTWS424YERXXL
- PRTWS424YERXXXL
- Model:
- S424
- S424
- S424
- S424
- S424
- S424
- S424
- S424
- S424
- S424
- S424
- S424
- Outfit applikationer:
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Industrial|Outdoor
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Køn:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Lukkestørrelse:
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Stof:
- 94% polyester, 6% elastane 160 g laminated to 100% polyester
- 94% polyester, 6% elastane 160 g laminated to 100% polyester
- 94% polyester, 6% elastane 160 g laminated to 100% polyester
- 94% polyester, 6% elastane 160 g laminated to 100% polyester
- 94% polyester, 6% elastane 160 g laminated to 100% polyester
- 94% polyester, 6% elastane 160 g laminated to 100% polyester
- 94% polyester, 6% elastane 160 g laminated to 100% polyester
- 94% polyester, 6% elastane 160 g laminated to 100% polyester
- 94% polyester, 6% elastane 160 g laminated to 100% polyester
- 94% polyester, 6% elastane 160 g laminated to 100% polyester
- 94% polyester, 6% elastane 160 g laminated to 100% polyester
- 94% polyester, 6% elastane 160 g laminated to 100% polyester
- Vægt:
- 310 g
- 310 g
- 310 g
- 310 g
- 310 g
- 310 g
- 310 g
- 310 g
- 310 g
- 310 g
- 310 g
- 310 g
Specifikationer
Product Family Options
Produktinformation
- ModelFarveTøj størrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.PRTWS424ORRSModelS424Outfit applikationerIndustrial|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof94% polyester, 6% elastane 160 g laminated to 100% polyesterVægt310 g - Specifikationer:
More
Specifications
Cat. No.PRTWS424ORRMModelS424Outfit applikationerIndustrial|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof94% polyester, 6% elastane 160 g laminated to 100% polyesterVægt310 g - Specifikationer:
More
Specifications
Cat. No.PRTWS424ORRLModelS424Outfit applikationerIndustrial|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof94% polyester, 6% elastane 160 g laminated to 100% polyesterVægt310 g - Specifikationer:
More
Specifications
Cat. No.PRTWS424ORRXLModelS424Outfit applikationerIndustrial|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof94% polyester, 6% elastane 160 g laminated to 100% polyesterVægt310 g - Specifikationer:
More
Specifications
Cat. No.PRTWS424ORRXXLModelS424Outfit applikationerIndustrial|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof94% polyester, 6% elastane 160 g laminated to 100% polyesterVægt310 g - Specifikationer:
More
Specifications
Cat. No.PRTWS424ORRXXXLModelS424Outfit applikationerIndustrial|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof94% polyester, 6% elastane 160 g laminated to 100% polyesterVægt310 g - Specifikationer:
More
Specifications
Cat. No.PRTWS424YERSModelS424Outfit applikationerIndustrial|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof94% polyester, 6% elastane 160 g laminated to 100% polyesterVægt310 g - Specifikationer:
More
Specifications
Cat. No.PRTWS424YERMModelS424Outfit applikationerIndustrial|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof94% polyester, 6% elastane 160 g laminated to 100% polyesterVægt310 g - Specifikationer:
More
Specifications
Cat. No.PRTWS424YERLModelS424Outfit applikationerIndustrial|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof94% polyester, 6% elastane 160 g laminated to 100% polyesterVægt310 g - Specifikationer:
More
Specifications
Cat. No.PRTWS424YERXLModelS424Outfit applikationerIndustrial|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof94% polyester, 6% elastane 160 g laminated to 100% polyesterVægt310 g - Specifikationer:
More
Specifications
Cat. No.PRTWS424YERXXLModelS424Outfit applikationerIndustrial|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof94% polyester, 6% elastane 160 g laminated to 100% polyesterVægt310 g - Specifikationer:
More
Specifications
Cat. No.PRTWS424YERXXXLModelS424Outfit applikationerIndustrial|OutdoorRisk areaRain (waterproof)KønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof94% polyester, 6% elastane 160 g laminated to 100% polyesterVægt310 g