Rainwear softshell jackets, TK53
Om denne vare
A contemporary detachable hooded jacket with a host of technical features, like pre-formed elbows and rubber cuff straps with 5 pockets for ample storage, phone pocket and zipped pockets.
- Micro polar fleece lined and bonded with shell
- Waterproof zips to prevent water penetration
- Internal pocket for safe storage
- Hook and loop cuffs for a secure fit
- Chin guard for added comfort and stability
Shell fabric: 94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar fleece 150 g 310 g.
Made of durable breathable, windproof and water resistant fabric.
7 tilgængelige muligheder nedenfor
Product Details & Documents
A contemporary detachable hooded jacket with a host of technical features, like pre-formed elbows and rubber cuff straps with 5 pockets for ample storage, phone pocket and zipped pockets.
- Micro polar fleece lined and bonded with shell
- Waterproof zips to prevent water penetration
- Internal pocket for safe storage
- Hook and loop cuffs for a secure fit
- Chin guard for added comfort and stability
Shell fabric: 94% polyester, 6% elastane 160 g laminated to 100% polyester micro polar fleece 150 g 310 g.
Made of durable breathable, windproof and water resistant fabric.
Specifications for Product Family
Specifikationer
- Catalog No:
- PRTWTK53BKRM
- PRTWTK53BKRL
- PRTWTK53BKRXL
- PRTWTK53BKRXXL
- PRTWTK53NARM
- PRTWTK53NARL
- PRTWTK53NARXL
- Model:
- TK53
- TK53
- TK53
- TK53
- TK53
- TK53
- TK53
- Outfit applikationer:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Køn:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Lukkestørrelse:
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Med Velcro®
- Stof:
- 94% polyester, 6% elastane
- 94% polyester, 6% elastane
- 94% polyester, 6% elastane
- 94% polyester, 6% elastane
- 94% polyester, 6% elastane
- 94% polyester, 6% elastane
- 94% polyester, 6% elastane
- Vægt:
- 160 g
- 160 g
- 160 g
- 160 g
- 160 g
- 160 g
- 160 g
Specifikationer
Product Family Options
Produktinformation
- ModelFarveTøj størrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.PRTWTK53BKRMModelTK53Outfit applikationerGeneral PurposeRisk areaRain (waterproof)KønUnisexLukkestørrelseMed Velcro®Stof94% polyester, 6% elastaneVægt160 g - Specifikationer:
More
Specifications
Cat. No.PRTWTK53BKRLModelTK53Outfit applikationerGeneral PurposeRisk areaRain (waterproof)KønUnisexLukkestørrelseMed Velcro®Stof94% polyester, 6% elastaneVægt160 g - Specifikationer:
More
Specifications
Cat. No.PRTWTK53BKRXLModelTK53Outfit applikationerGeneral PurposeRisk areaRain (waterproof)KønUnisexLukkestørrelseMed Velcro®Stof94% polyester, 6% elastaneVægt160 g - Specifikationer:
More
Specifications
Cat. No.PRTWTK53BKRXXLModelTK53Outfit applikationerGeneral PurposeRisk areaRain (waterproof)KønUnisexLukkestørrelseMed Velcro®Stof94% polyester, 6% elastaneVægt160 g - Specifikationer:
More
Specifications
Cat. No.PRTWTK53NARMModelTK53Outfit applikationerGeneral PurposeRisk areaRain (waterproof)KønUnisexLukkestørrelseMed Velcro®Stof94% polyester, 6% elastaneVægt160 g - Specifikationer:
More
Specifications
Cat. No.PRTWTK53NARLModelTK53Outfit applikationerGeneral PurposeRisk areaRain (waterproof)KønUnisexLukkestørrelseMed Velcro®Stof94% polyester, 6% elastaneVægt160 g - Specifikationer:
More
Specifications
Cat. No.PRTWTK53NARXLModelTK53Outfit applikationerGeneral PurposeRisk areaRain (waterproof)KønUnisexLukkestørrelseMed Velcro®Stof94% polyester, 6% elastaneVægt160 g