Winter overalls, S585
Om denne vare
These are full waterproof protection and outstanding durability are key features of this winter padded overalls. The contemporary design is complemented by a high quality finish and close attention to detail throughout.
- Abrasion resistant oxford polyester
- Taped seams to provide additional protection
- Quilt lined for thermal insulation
- Five pockets for ample storage
- Phone pocket
- Concealed stud and zip fastening
- Two-way zip for quick and easy access
- Fully elasticated waistband for ultimate wearer comfort
Key features includes: Reflective tape for added visibility, reinforced knee patches for optimum strength, multiple pockets for storage and zipped ankles for easy fitting.
Weight (polyester lining fabric): 60 g/m²
Weight (polyester filling fabric): 170 g/m²
Weight (Shell fabric): 190 g/m²
Certifications: CE certified, EN 342 0,341m². K/W (B), 2, X, EN 343:2019 Class 3:1 X WP 15,000 mm
10 tilgængelige muligheder nedenfor
Product Details & Documents
These are full waterproof protection and outstanding durability are key features of this winter padded overalls. The contemporary design is complemented by a high quality finish and close attention to detail throughout.
- Abrasion resistant oxford polyester
- Taped seams to provide additional protection
- Quilt lined for thermal insulation
- Five pockets for ample storage
- Phone pocket
- Concealed stud and zip fastening
- Two-way zip for quick and easy access
- Fully elasticated waistband for ultimate wearer comfort
Key features includes: Reflective tape for added visibility, reinforced knee patches for optimum strength, multiple pockets for storage and zipped ankles for easy fitting.
Weight (polyester lining fabric): 60 g/m²
Weight (polyester filling fabric): 170 g/m²
Weight (Shell fabric): 190 g/m²
Certifications: CE certified, EN 342 0,341m². K/W (B), 2, X, EN 343:2019 Class 3:1 X WP 15,000 mm
Specifications for Product Family
Specifikationer
- Catalog No:
- PRTWS585GRRS
- PRTWS585NARS
- PRTWS585GRRM
- PRTWS585NARM
- PRTWS585NARL
- PRTWS585GRRL
- PRTWS585GRRXL
- PRTWS585NARXL
- PRTWS585GRRXXL
- PRTWS585NARXXL
- Model:
- Winter overalls
- Winter overalls
- Winter overalls
- Winter overalls
- Winter overalls
- Winter overalls
- Winter overalls
- Winter overalls
- Winter overalls
- Winter overalls
- Outfit applikationer:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Risk area:
- Rain (waterproof)|Cold
- Rain (waterproof)|Cold
- Rain (waterproof)|Cold
- Rain (waterproof)|Cold
- Rain (waterproof)|Cold
- Rain (waterproof)|Cold
- Rain (waterproof)|Cold
- Rain (waterproof)|Cold
- Rain (waterproof)|Cold
- Rain (waterproof)|Cold
- Køn:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Lukkestørrelse:
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Stof:
- 100% polyester, PU coated
- 100% polyester, PU coated
- 100% polyester, PU coated
- 100% polyester, PU coated
- 100% polyester, PU coated
- 100% polyester, PU coated
- 100% polyester, PU coated
- 100% polyester, PU coated
- 100% polyester, PU coated
- 100% polyester, PU coated
- Vægt:
- 60 g/m² + 170 g/m² + 190 g/m²
- 60 g/m² + 170 g/m² + 190 g/m²
- 60 g/m² + 170 g/m² + 190 g/m²
- 60 g/m² + 170 g/m² + 190 g/m²
- 60 g/m² + 170 g/m² + 190 g/m²
- 60 g/m² + 170 g/m² + 190 g/m²
- 60 g/m² + 170 g/m² + 190 g/m²
- 60 g/m² + 170 g/m² + 190 g/m²
- 60 g/m² + 170 g/m² + 190 g/m²
- 60 g/m² + 170 g/m² + 190 g/m²
Specifikationer
Product Family Options
Produktinformation
- ModelFarveTøj størrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.PRTWS585GRRSModelWinter overallsOutfit applikationerGeneral PurposeRisk areaRain (waterproof)|ColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyester, PU coatedVægt60 g/m² + 170 g/m² + 190 g/m² - Specifikationer:
More
Specifications
Cat. No.PRTWS585NARSModelWinter overallsOutfit applikationerGeneral PurposeRisk areaRain (waterproof)|ColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyester, PU coatedVægt60 g/m² + 170 g/m² + 190 g/m² - Specifikationer:
More
Specifications
Cat. No.PRTWS585GRRMModelWinter overallsOutfit applikationerGeneral PurposeRisk areaRain (waterproof)|ColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyester, PU coatedVægt60 g/m² + 170 g/m² + 190 g/m² - Specifikationer:
More
Specifications
Cat. No.PRTWS585NARMModelWinter overallsOutfit applikationerGeneral PurposeRisk areaRain (waterproof)|ColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyester, PU coatedVægt60 g/m² + 170 g/m² + 190 g/m² - Specifikationer:
More
Specifications
Cat. No.PRTWS585NARLModelWinter overallsOutfit applikationerGeneral PurposeRisk areaRain (waterproof)|ColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyester, PU coatedVægt60 g/m² + 170 g/m² + 190 g/m² - Specifikationer:
More
Specifications
Cat. No.PRTWS585GRRLModelWinter overallsOutfit applikationerGeneral PurposeRisk areaRain (waterproof)|ColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyester, PU coatedVægt60 g/m² + 170 g/m² + 190 g/m² - Specifikationer:
More
Specifications
Cat. No.PRTWS585GRRXLModelWinter overallsOutfit applikationerGeneral PurposeRisk areaRain (waterproof)|ColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyester, PU coatedVægt60 g/m² + 170 g/m² + 190 g/m² - Specifikationer:
More
Specifications
Cat. No.PRTWS585NARXLModelWinter overallsOutfit applikationerGeneral PurposeRisk areaRain (waterproof)|ColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyester, PU coatedVægt60 g/m² + 170 g/m² + 190 g/m² - Specifikationer:
More
Specifications
Cat. No.PRTWS585GRRXXLModelWinter overallsOutfit applikationerGeneral PurposeRisk areaRain (waterproof)|ColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyester, PU coatedVægt60 g/m² + 170 g/m² + 190 g/m² - Specifikationer:
More
Specifications
Cat. No.PRTWS585NARXXLModelWinter overallsOutfit applikationerGeneral PurposeRisk areaRain (waterproof)|ColdKønUnisexSleeve or leg lengthLongLukkestørrelseMed lynlåsStof100% polyester, PU coatedVægt60 g/m² + 170 g/m² + 190 g/m²