LED ring light, VisiLED, BF S40-75
VisiLED ring light, BF S40-75, int.Ø 66 mm, ext.Ø 92 mm, working distance 55 - 155 mm, max. 110 klx (85 mm)
Katalog # SCOC400110
Leverandør: SCHOTT AG Lighting and imaging
LED ring light, VisiLED, BF S40-75
Katalog # SCOC400110
Leverandør: SCHOTT AG Lighting and imaging
Gå til:
Product Details & Documents
The VisiLED ringlight S40-75 is the successor of the S40-55, which has been discontinued, and comes with a completely new design.
- Slim housing; fits between two lenses for usage with objective revolver
- 110 klux (WD 85 mm)
- Extra homogenous illumination
- Lifetime is typically 50000 h by actively controlling the LED‘s temperature
Designed for use with controller MC 1100 or MC 1500.
Bestillingsoplysninger: Controller MC 1100 or MC 1500 required.
Documents
Ring Lights, EasyLED
Videoer
VisiLED Series Professional Illumination for Stereo Microscopy
Produktlitteratur
Specifikationer
- Beskrivelse:VisiLED ring light, BF S40-75, int.Ø 66 mm, ext.Ø 92 mm, working distance 55 - 155 mm, max. 110 klx (85 mm)