Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Eusolex® 232

Eusolex® 232

1 kg
Catalog # 1.05372.1001
Supplier:   Merck
Eusolex® 232
Catalog # 1.05372.1001
Supplier:  Merck
CAS Number:  27503-81-7
Eusolex® 232

1 kg

CAS Number:  27503-81-7
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents

Organic UV Filter

Eusolex® 232 is a highly effective UV-B filter for the water phase. It's a white powder with excellent protection properties and clearly soluble in water after having completely neutralized it with bases to pH > 7 (e.g. by Sodium Hydroxide, TEA or TRIS). Formulating of clear aqueous gels is possible. Eusolex® 232 is photostable and approved worldwide. Together with oil soluble, sunscreens synergistic SPF's can be achieved.

INCI name : Phenylbenzimidazole Sulfonic Acid

  • CAS Number:27503-81-7
  • Formula:C₁₃H₁₀N₂O₃S
  • Storage Temperature:Stue temperatur

Documents

Specifications

Specifications

  • Pakn:
    1 kg

Specifications