Polyester labels for M611 and BMP™61 printers, type B-490 and B-492, Freezerbondz™
Katalog # BRDY150870
Leverandør: Brady
Specifikationer
- Model:B-492
- Beskrivelse:Freezerbondz™ labels
- Dimensioner:12,70H×25,40W mm
- Materiale:Polyester
- Seq:05
- Pakn:1 Roll
- Klæbestof:Permanent akryl
Specifikationer
Om denne vare
Die-cut labels and continuous tapes are made from Freezerbondz™ polyester, with a permanent acrylic adhesive and matte finish.
- Die-cut labels are supplied on a roll
- Continuous tape is not cut to length; it is cut by the printer
- Label thickness is 0,065mm (B-490) and 0,081 mm (B-492)
- Maximum service temperature: 130 °C (B-490) and 110 °C (B-492)
- Minimum service temperature: –196 °C
- Ribbon series: R4300, R6400
Durable Freezerbondz™ polyester B-490 and B-492 labels are designed for use in laboratory identification applications, such as vials, centrifuge, tubes, test tubes, straws, and slides.