Orders LinkContactUsLinkComponent
HyClone Cell Boost™ 7b supplement
HyClone Cell Boost™ 7b supplement
HyClone Cell Boost™ 7b supplement
Certifikater
HyClone Cell Boost™ 7b supplement
5 tilgængelige muligheder nedenfor
Product Details & Documents
Supplements cell culture with concentrated amino acids. Manufactured to meet cGMP manufacturing standards and QC specifications.
  • Designed to support modern manufacturing processes, Cell Boost 7b supplement is a concentrated solution of amino acids with an alkaline pHSupplement is chemically defined, animal-derived component-free (ADCF), do not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
  • Use Cell Boost 7a and 7b together to achieve high yields of mAbs and other proteins in CHO fed-batch processes
  • For convenience, this cell culture supplement is available as a ready-to-use liquid feed solution in addition to a dry powder format
  • Non sterile
Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture. HyClone Cell Boost 7a and Cell Boost 7b supplements are intended for use in combination with HyClone ActiPro and ActiSM media as part of the ActiPro cell culture system formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines. Cell Boost 7b is available as a convenient ready-to-use liquid feed solution or as a dry powder feed to be hydrated before use.
Specifications for Product Family
Specifikationer
Specifikationer
  • Catalog No:
  • SH31120.01
  • SH31120.02
  • SH31120.03
  • SH31027.01
  • SH31027.07
  • Shelf life:
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • Opbevaring betingelser:
  • Away from light and moisture
  • Away from light and moisture
  • Away from light and moisture
  • Away from light and moisture
  • Away from light and moisture
  • Opbevaringstemperatur:
  • 2…8 °C
  • 2…8 °C
  • 2…8 °C
  • 2…8 °C
  • 2…8 °C
Specifikationer
Product Family Options
Produktinformation
  • Form
    Størrelse
    Pakn
    Tilgængelighed
    Price
  • SH31120.01
    Cell Boost™ 7b
    Cell Boost™ 7b
    Liquid
    100 ml
    100 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mL
    1 * 100 mL - 635,00 kr
    635,00 kr
    1 * 100 mL
       635,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    SH31120.01
    Shelf life
    18 months
    Opbevaring betingelser
    Away from light and moisture
    Opbevaringstemperatur
    2…8 °C
  • SH31120.02
    Cell Boost™ 7b
    Cell Boost™ 7b
    Liquid
    500 ml
    500 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 mL
    1 * 500 mL - 2.650,00 kr
    2.650,00 kr
    1 * 500 mL
       2.650,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    SH31120.02
    Shelf life
    18 months
    Opbevaring betingelser
    Away from light and moisture
    Opbevaringstemperatur
    2…8 °C
  • SH31120.03
    Cell Boost™ 7b
    Cell Boost™ 7b
    Liquid
    1000 ml
    1.000 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 mL
    1 * 1.000 mL - 4.090,00 kr
    4.090,00 kr
    1 * 1.000 mL
       4.090,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    SH31120.03
    Shelf life
    18 months
    Opbevaring betingelser
    Away from light and moisture
    Opbevaringstemperatur
    2…8 °C
  • SH31027.01
    Cell Boost™ 7b
    Cell Boost™ 7b
    Powder
    500 ml
    1
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.000,00 kr
    2.000,00 kr
    1 * 1 Items
       2.000,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    SH31027.01
    Shelf life
    18 months
    Opbevaring betingelser
    Away from light and moisture
    Opbevaringstemperatur
    2…8 °C
  • SH31027.07
    Cell Boost™ 7b
    Cell Boost™ 7b
    Powder
    1 L
    1 L
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 L
    1 * 1 L - 3.580,00 kr
    3.580,00 kr
    1 * 1 L
       3.580,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    SH31027.07
    Shelf life
    18 months
    Opbevaring betingelser
    Away from light and moisture
    Opbevaringstemperatur
    2…8 °C