Tributyltin chloride [D27] 100 µg/ml in methylene chloride [D2] ≥98%
Catalog # CAMBDLM-7136-1.2
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pakn:1,2 mL
- Seq:2080
Specifications
About this item
:
:352.67 g/mol
CAS Number:1257647-76-9
Formula:C₁₂D₂₇ClSn
MDL Number:MFCD03428112