Order Entry
Denmark
ContactUsLinkComponent
Mono-(2-ethyl-5-hydroxyhexyl)phtalate (DEHP Metabolite IX) [13C4]
Mono-(2-ethyl-5-hydroxyhexyl)phtalate (DEHP Metabolite IX) [13C4]
Catalog # CAMBCLM-6641-1.2
CAS Number:  
Mono-(2-ethyl-5-hydroxyhexyl)phtalate (DEHP Metabolite IX) [13C4]
Catalog # CAMBCLM-6641-1.2
CAS Number:  

Specifications

  • Pakn:
    1,2 mL
  • Seq:
    2080

Specifications

About this item

 :
 :298.31 g/mol
 :40321-99-1
Formula:C₁₂*C₄H₂₂O₅