Order Entry
Denmark
ContactUsLinkComponent
Carbaryl(1-naphthyl methylcarbamate) [ring-13C6] 100 µg/ml in n-nonane
Flammable
Irritant
Health
Carbaryl(1-naphthyl methylcarbamate) [ring-13C6] 100 µg/ml in n-nonane
Catalog # CAMBCLM-4682-1.2
CAS Number:  
Flammable
Irritant
Health
Carbaryl(1-naphthyl methylcarbamate) [ring-13C6] 100 µg/ml in n-nonane
Catalog # CAMBCLM-4682-1.2
CAS Number:  

Specifications

  • Pakn:
    1,2 mL
  • Seq:
    2080

Specifications

About this item

 :
 :207.18 g/mol
 :GHS02
 :GHS07
 :GHS08
CAS Number:2483735-95-9
Formula:*C₆C₆H₁₁NO₂