Orders LinkContactUsLinkComponent
Spill kaddie
Spill kaddie
Spill kaddie, chemical, 59 L
Katalog # 121-0198
Leverandør:   Brady
Spill kaddie
Katalog # 121-0198
Leverandør:  Brady
Spill kaddie
Spill kaddie
Spill kaddie, chemical, 59 L
Price
6.110,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
iThis is a compact wheeled spill kit for fast spill response or maintenance jobs - for chemicals and hazardous liquids.
  • Double doors provide easy access to products
  • Yellow in colour
Interior shelves and compartments keep sorbents organised. Cart protects sorbents against dirt, moisture and damage. Fits easily through narrow aisles and doorways. Wheels allow excellent mobility both indoors and outdoors.
Leveringsinformation: Includes 1 double perforated roll (380×2200 mm), 2 socs (Ø 76×1220 mm), 1 soc (Ø 76×2440 mm), 6 pillows (250×250 mm), 1 pair of goggles, 1 pair of gloves, 2 disposal bags.
Specifikationer
  • Beskrivelse:
    Spill kaddie, chemical
  • Sugeevne:
    59 L