Labels for BBP33 printers, type B-486
Supplier: Brady
Jump to:
About this item
Metallised polyester labels for product labelling, asset tracking and general industrial ID.
- Matte, light gray material has a permanent rubber-based adhesive and is thermal transfer printable
- Ideal for serial number plates and rating plates with name-plate quality and metal-looking finish
- Service temperature from –40 to +120 °C
: UL approved, CSA accepted.
9 Options Available Below
Product Details & Documents
Metallised polyester labels for product labelling, asset tracking and general industrial ID.
- Matte, light gray material has a permanent rubber-based adhesive and is thermal transfer printable
- Ideal for serial number plates and rating plates with name-plate quality and metal-looking finish
- Service temperature from –40 to +120 °C
: UL approved, CSA accepted.
Product Family Options
Product Information
- BeskrivelseModelMaterialeKlæbestofPrinting methodFarvePrinterDimensionerAvailabilityPrice
- Metalised printer labelsB-486PolyesterPermanent, gummibaseretThermal transfersølvfarvetBBP3325,4×12,7 mmLog in for availability1 * 250 Itemskr1,500.00
- Metalised printer labelsB-486PolyesterPermanent, gummibaseretThermal transfersølvfarvetBBP3325,4×12,7 mmLog in for availability1 * 2.500 Itemskr2,030.00
- Metalised printer labelsB-486PolyesterPermanent, gummibaseretThermal transfersølvfarvetBBP3350,8×12,7 mmLog in for availability1 * 2.500 Itemskr1,170.00
- Metalised printer labelsB-486PolyesterPermanent, gummibaseretThermal transfersølvfarvetBBP3350,8×25,4 mmLog in for availability1 * 1.000 Itemskr1,650.00
- Metalised printer labelsB-486PolyesterPermanent, gummibaseretThermal transfersølvfarvetBBP3369,9×31,8 mmLog in for availability1 * 500 Itemskr2,230.00
- Metalised printer labelsB-486PolyesterPermanent, gummibaseretThermal transfersølvfarvetBBP3376,2×25,4 mmLog in for availability1 * 5.000 Itemskr2,320.00
- Metalised printer labelsB-486PolyesterPermanent, gummibaseretThermal transfersølvfarvetBBP3376,2×50,8 mmLog in for availability1 * 500 Itemskr1,440.00
- Metalised printer labelsB-486PolyesterPermanent, gummibaseretThermal transfersølvfarvetBBP33101,6×76,2 mmLog in for availability1 * 1.300 Itemskr1,870.00
- Metalised printer labelsB-486PolyesterPermanent, gummibaseretThermal transfersølvfarvetBBP33101,6×101,6 mmLog in for availability1 * 1.300 Itemskr1,320.00