Orders LinkContactUsLinkComponent
SlideMop™ Mops
Leverandør:   MICRONOVA
SlideMop™ Mops
Leverandør:  MICRONOVA
Certifikater
SlideMop™ Mops
SlideMop™ Mops
5 tilgængelige muligheder nedenfor
Product Details & Documents
A simple, effective mop system featuring sturdy, collapsible plates that easily slide into the quilted polyester mop cover for use in traditional 'dip and wring' methods, or as a 'saturation' method where liquid disposal and/or space-saving is a factor.
  • Lightweight, economical design for use throughout the facility
  • Quilted polyester mop covers with polyester pad interiors for disinfectant dispersal
  • Choice of stainless steel or plastic frame
  • Swivel joint ensures correct lift and pull for figure eight cleaning techniques
  • Use with W-94 or W-92 wringer
  • Available gamma irradiated: 25 kGy minimum dose
  • Good chemical resistance
Designed for use with most threaded plastic and aluminum handles and can be used with the quick disconnect stainless steel handles and ACME adapter.
 : Manufactured for use in controlled environments. ISO 5 through to 9 cleanrooms. ISO 9001 certified manufacturing facility.
Product Family Options
Covers, SlideMop™
Produktinformation
  • Dimensioner
    L×B
    Tilgængelighed
    Price
  • 129-1599
    SlideMop™ irradiated covers, polyester
    SlideMop™ irradiated covers, polyester
    400 mm
    406×140 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 24 Items
    1 * 24 Items - 3.000,00 kr
    3.000,00 kr
    1 * 24 Items
       3.000,00 kr
  • MICRMZCR63-16IR
    SlideMop™ irradiated covers
    SlideMop™ irradiated covers
    400 mm
    406×140 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 24 Items
    1 * 24 Items - 3.000,00 kr
    3.000,00 kr
    1 * 24 Items
       3.000,00 kr
  • 129-1598
    SlideMop™ covers, polyester
    SlideMop™ covers, polyester
    400 mm
    406×140 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 24 Items
    1 * 24 Items - 2.560,00 kr
    2.560,00 kr
    1 * 24 Items
       2.560,00 kr
Frame, SlideMop™
Produktinformation
  • Dimensioner
    L×B
    Tilgængelighed
    Price
  • MICRQPSM-16
    SlideMop™ stainless steel frame
    SlideMop™ stainless steel frame
    400 mm
    406×127 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 5.250,00 kr
    5.250,00 kr
    1 * 1 Items
       5.250,00 kr
  • 129-1606
    SlideMop™ plastic frame, polypropylene
    SlideMop™ plastic frame, polypropylene
    400 mm
    406×127 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 738,00 kr
    738,00 kr
    1 * 1 Items
       738,00 kr