Kaliumpolysulfider
Glass bottle for solids, 1 kg
Katalog # ROTH8010.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Potassium polysulphide, Liver of sulphur, Sulphurated potash
- CAS nummer:37199-66-9
- Formel:K₂(S)n
- MDL nummer:MFCD00148074
- Molecular Weight:110.26 g/mol
- Smeltende Pt:471 °C
- Tæthed:1,65 g/cm³ (20 °C)
- Opbevaringstemperatur:Køleskab
- EINECS:253-390-1
- FN:1382
- ADR:4.2,II
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg