Methyl dihydroabietate
25 g
Katalog # TCIAD1088-25G
Leverandør: TCI
Gå til:
Product Details & Documents
Synonymer: Dihydroabietic Acid Methyl Ester
- CAS nummer:30968-45-7
- Formel:C₂₁H₃₄O₂
- MDL nummer:MFCD00466887
- Molecular Weight:318.50 g/mol
- Flash Pt:183°C
Specifikationer
Specifikationstestresultater
- Appearance:Light yellow to Brown clear liquid
- Purity (GC):many component mixture
Specifikationer
- Pakn:25 g



