Orders LinkContactUsLinkComponent
Streptavidin, Cy5®
Streptavidin, Cy5®
1 mg
Katalog # PA45001
Leverandør:   Cytiva
Streptavidin, Cy5®
Katalog # PA45001
Leverandør:  Cytiva
CAS nummer:  
Streptavidin, Cy5®
Streptavidin, Cy5®
1 mg
CAS nummer:  
Price
2.250,00 kr
Log ind for tilgængelighed
1 * 1 mg
Product Details & Documents
Streptavidin-fluor conjugates suitable for use in the fluorescent analysis of biotinylated samples.
  • High specificity reagents
This product is supplied as a lyophilized solid in phosphate-buffered saline (0.02 M potassium phosphate, 0.15 M NaCl) pH 7.2, containing 1%(w/v) bovine serum albumin and 0.1% (w/v) sodium azide. Reconstitute with 1.0 ml of deionized water, dispense into suitable aliquots and freeze. Avoid repeated freeze-thaw cycles.
Specifikationer
Specifikationer
  • Pakn:
    1 mg
  • Cat. No.:
    PA45001
  • Konjugation:
    Cy5®
  • Form:
    Solid
  • Protein/peptide name:
    Streptavidin
  • Opløselighed:
    Soluble in cold water and hot water
  • Renhed:
    This product was prepared from electrophoretically isolated and purified streptavidin from Streptomyces Avidinii. A single precipitin arc was observed against anti-streptavidin when assayed by immunoelectrophoresis
  • Opbevaring betingelser:
    2 - 8 °C
  • Farve:
    Blue