ContactUsLinkComponent
Phosphat 1000 mg/l i vand (af KH2PO4) Certipur®, Supelco®
Phosphat 1000 mg/l i vand (af KH2PO4) Certipur®, Supelco®
Catalog # 1.19898.0500
Supplier:  Merck
CAS Number:  
Phosphat 1000 mg/l i vand (af KH2PO4) Certipur®, Supelco®
Catalog # 1.19898.0500
Supplier:  Merck
CAS Number:  
Specifications
  • Koncentration:
    1000 mg/L
  • Pakning:
    Plastic bottle
  • Pakn:
    500 mL
  • Beskrivelse:
    Fosfat standardopløsning
  • M-Clarity quality level:
    MQ100