Natriumhypochlorit solution (5% Cl₂) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Plastic bottle, 1 L
Leverandør: Avantor
Natriumhypochlorit solution (5% Cl₂) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Katalog # 7668.1000
Leverandør: Avantor
CAS nummer: 7681-52-9
Gå til:
Product Details & Documents
- CAS nummer:7681-52-9
- Formel:ClNaO
- MDL nummer:MFCD00011120
- Molecular Weight:74.44 g/mol
- Kogende Pt:111 °C (1013 hPa)
- Smeltende Pt:–30…–20 °C
- Tæthed:1,22…1.25 g/cm³ (20 °C)
- Opbevaringstemperatur:Køleskab
- EINECS:231-668-3
- FN:1791
- ADR:8,II
Documents
Specifikationer
Specifikationstestresultater
- Active Chlorine:0.055 - 0.075%
- pH at 25 °C:min. 11.5
Specifikationer
- Pakning:Plastic bottle
- Pakn:1 L