Orcein
Glass bottle for solids, 5 g
Catalog # SERA31178.01
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Orcein (from synthetic), Natural Red 28
- CAS Number:1400-62-0
- MDL Number:MFCD00062310
- EINECS:215-750-6
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:5 g