Saponin
Supplier: VWR Chemicals
About this item
- CAS Number:8047-15-2
- MDL Number:MFCD00081981
- Density:1,015…1.02 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:232-462-6
2 Options Available Below
Product Details & Documents
- CAS Number:8047-15-2
- MDL Number:MFCD00081981
- Density:1,015…1.02 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:232-462-6
Documents
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 20 L Plastic bucket for solids UN Z (ZPACK13247020)
Packaging Information
440914Y_SAPONIN BIOCHEMICAL_BSE_REV0.pdf
BSE/TSE Certificates
Specifications for Product Family
Specification Test Results
- Identification:Passes test
- pH (5 %):4 - 7
- Surface tension (20°C; 0,1%; water):30 - 60 mN/m
Product Family Options
Product Information