ContactUsLinkComponent
SDS-PAGE Sample Buffer, 2X, (2-Mercaptoethanol-free) for Electrophoresis
SDS-PAGE Sample Buffer, 2X, (2-Mercaptoethanol-free) for Electrophoresis
Catalog # TCIAB5834-25ML
Supplier:  TCI
CAS Number:  
SDS-PAGE Sample Buffer, 2X, (2-Mercaptoethanol-free) for Electrophoresis
Catalog # TCIAB5834-25ML
Supplier:  TCI
CAS Number:  
Specifications
  • Pakn:
    25 mL
  • Beskrivelse:
    SDS-PAGE Sample Buffer, 2X
Specifications
About this item
SDS-PAGE Sample Buffer is specially formulated for protein sample preparation to based on the Laemmli Method of SDS-PAGE system.
This product contains 4% SDS, 20% glycerol, 0,01% bromophenol blue and 0,125 M Tris HCl, pH approx. 6,8. Does not contain 2-Mercaptoethanol.
Caution: Please use after bringing back to normal temperature.