Orders LinkContactUsLinkComponent
VWR®, Wire baskets with two handles
VWR®, Wire baskets with two handles
Leverandør:   VWR Collection
VWR®, Wire baskets with two handles
Leverandør:  VWR Collection
Certifikater
VWR®, Wire baskets with two handles
VWR®, Wire baskets with two handles
3 tilgængelige muligheder nedenfor
Product Details & Documents
These wire baskets are made from rust-proof, electrolytically polished stainless steel.
  • Very good chemical and temperature resistance
  • Two flip handles
  • Base frame is stackable
Suitable for sterilisation of laboratory utensils.
Product Family Options
Produktinformation
  • B×D×H
    Tilgængelighed
    Price
  • 240×250×50 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.180,00 kr
    1.180,00 kr
    1 * 1 Items
       1.180,00 kr
  • 480×250×60 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.560,00 kr
    1.560,00 kr
    1 * 1 Items
       1.560,00 kr
  • 540×255×60 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.250,00 kr
    2.250,00 kr
    1 * 1 Items
       2.250,00 kr