Pipette workstation
Pipette workstation, 4 pipettes, multiple size tips, klar, 244×165×246 mm
Katalog # HEATHS234631
Leverandør: HEATHROW
Gå til:
Product Details & Documents
Keep pipette and disposable tips together in one convenient stand.
- Holds up to four pipettes on top shelf
- Dual bins hold multiple size tips
- Self-closing lid provides dust-free storage
- Oversized lip allows for easy access when wearing gloves
Acrylic workstation is opaque on sides with clear front and bin lids for easy content identification.
Specifikationer
- Til:4 pipettes, multiple size tips
- Farve:klar
- Beskrivelse:Pipette workstation
- B×D×H:244×165×246 mm