Orders LinkContactUsLinkComponent
Calcium assay kit
Calcium assay kit
Calcium Assay Kit, 192 Tests
Katalog # CAYM701220-192
Leverandør:   Cayman Chemical
Calcium assay kit
Katalog # CAYM701220-192
Leverandør:  Cayman Chemical
CAS nummer:  
Calcium assay kit
Calcium Assay Kit, 192 Tests
CAS nummer:  
Price
3.330,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
For the determination of total calcium concentration in a variety of biological samples, as well as tissue homogenates and cell lysates.
  • Measure calcium in plasma, serum, urine, tissue homogenates, and cell lysates
  • Assay 40 samples in duplicate
  • Measure calcium down to 0,25 mg/dl
  • Plate-based colorimetric measurement (570 to 590 nm)
Specifikationer
Specifikationer
  • Pakn:
    192 Tests
  • Beskrivelse:
    Calcium Assay Kit
  • Cat. No.:
    CAYM701220-192
  • Detektionsgrænse:
    0.25 mg/dl