Orders LinkContactUsLinkComponent
Coated filter, ORBO™ 80
Coated filter, ORBO™ 80
37 mm
Katalog # 20811.
Leverandør:   Merck
Coated filter, ORBO™ 80
Katalog # 20811.
Leverandør:  Merck
Coated filter, ORBO™ 80
37 mm
Price
2.030,00 kr
Log ind for tilgængelighed
1 * 25 Items
Product Details & Documents
ORBO™ 80 is a glass fiber filter coated with 1-(2-pyridyl)piperazine.
  • No cassettes
It is used for sampling diisocyanates in air.
Specifikationer
  • Størrelse:
    37 mm