Orders LinkContactUsLinkComponent
TISAB II Solution, Supelco®
TISAB II Solution, Supelco®
TISAB II solution, Plastic bottle, 500 mL
Katalog # 89466-500ML
Leverandør:   Merck
TISAB II Solution, Supelco®
Katalog # 89466-500ML
Leverandør:  Merck
CAS nummer:  
TISAB II Solution, Supelco®
TISAB II solution, Plastic bottle, 500 mL
CAS nummer:  
Price
kr874.00
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
TISAB, in general, is mixed with samples, as it helps in maintaining the ionic strength and pH constant. It is mainly used to decomplex metal-F complexes.
TISAB II can be prepared using NaCl with glacial acetic acid, 1,2-Cyclohexylenedinitrilotetraacetic acid and NaOH. It also has high concentrations of cation chelators that hydrolyse fluoride-ion pairs to form fluoride ions.
TISAB II has been used to determine total fluoride concentration using fluoride ion sensitive electrode from dental varnish.
Documents
Specifikationer
Specifikationer
  • Pakning:
    Plastic bottle
  • Pakn:
    500 mL
  • Beskrivelse:
    TISAB II solution
  • Cat. No.:
    89466-500ML
  • M-Clarity quality level:
    MQ100
Specifikationer