Trousers, PP
Leverandør: HUBEI ORIENT INTL TRADING
Om denne vare
These disposable PP trousers are ideal for use in combination with PP lab coats.
- Breathable and lightweight fabric
- Easy to put on
- Elasticated waist and ankles
Weight: 40 g/m²
2 tilgængelige muligheder nedenfor
Product Details & Documents
These disposable PP trousers are ideal for use in combination with PP lab coats.
- Breathable and lightweight fabric
- Easy to put on
- Elasticated waist and ankles
Weight: 40 g/m²
Specifications for Product Family
Specifikationer
- Catalog No:
- HOIT109978
- HOIT145000
- Lommer:
- uden lommer
- uden lommer
- Køn:
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Outfit applikationer:
- General Purpose|Medical|Laboratory|Industrial
- General Purpose|Medical|Laboratory|Industrial
- Vægt:
- 40 g/m²
- 40 g/m²
Specifikationer
Product Family Options
Produktinformation
- FarveTøj størrelseTilgængelighedPrice
- HOIT10997810 pieces/bag; 10 bags/caseSpecifikationer:
More
Specifications
Cat. No.HOIT109978Lommeruden lommerKønUnisexSleeve or leg lengthLongOutfit applikationerGeneral Purpose|Medical|Laboratory|IndustrialVægt40 g/m² - HOIT14500010 pieces/bag; 10 bags/caseSpecifikationer:
More
Specifications
Cat. No.HOIT145000Lommeruden lommerKønUnisexSleeve or leg lengthLongOutfit applikationerGeneral Purpose|Medical|Laboratory|IndustrialVægt40 g/m²