Trace Metals 1-WP, certified reference material, Supelco®
Trace Metals 1-WP, 5% HNO₃, Ampoule, 20 mL
Katalog # QC1132-20ML
Leverandør: Merck
Trace Metals 1-WP, certified reference material, Supelco®
Katalog # QC1132-20ML
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
Analytes:
Aluminium; Arsenic; Beryllium;
Cadmium; Chromium; Cobalt; Copper;
Iron; Lead; Lithium;
Manganese; Mercury; Nickel;
Selenium; Vanadium; Zinc
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Specifikationer
Specifikationer
- Matrice:5% HNO₃
- Pakning:Ampoule
- Pakn:20 mL
- Beskrivelse:Trace Metals 1-WP